EMAIL THIS PAGE TO A FRIEND
Carbohydrate polymers
Production of starch nanoparticles by dissolution and non-solvent precipitation for use in food-grade Pickering emulsions.
PMID 27987962
Abstract
The aim of this study was to investigate non-solvent precipitation of starch to produce nanoparticles that could be used in Pickering emulsions. The material used was waxy maize, modified with octenyl succinic anhydride. Different methods of non-solvent precipitation were investigated, and a method based on direct mixing of an 8% starch solution and ethanol (ratio 1:1) was found to produce the smallest particles. The particle size was measured using AFM and AF4, and was found to be in the range 100-200nm. However, both larger particles and aggregates of nanoparticles were observed. The emulsion produced using the precipitated starch particles had a droplet size that between 0.5 and 45μm, compared to emulsions produced from waxy maize granules, in which had a size of 10-100μm. The drop in size contributed to increased stability against creaming. The amount of starch used for emulsion stabilization could also be substantially reduced.
Related Materials