EMAIL THIS PAGE TO A FRIEND
ACS applied materials & interfaces
Na2Ti3O7 Nanoplatelets and Nanosheets Derived from a Modified Exfoliation Process for Use as a High-Capacity Sodium-Ion Negative Electrode.
PMID 27996248
Abstract
The increasing interest in Na-ion batteries (NIBs) can be traced to sodium abundance, its low cost compared to lithium, and its intercalation chemistry being similar to that of lithium. We report that the electrochemical properties of a promising negative electrode material, Na2Ti3O7, are improved by exfoliating its layered structure and forming 2D nanoscale morphologies, nanoplatelets, and nanosheets. Exfoliation of Na2Ti3O7 was carried out by controlling the amount of proton exchange for Na(+) and then proceeding with the intercalation of larger cations such as methylammonium and propylammonium. An optimized mixture of nanoplatelets and nanosheets exhibited the best electrochemical performance in terms of high capacities in the range of 100-150 mA h g(-1) at high rates with stable cycling over several hundred cycles. These properties far exceed those of the corresponding bulk material, which is characterized by slow charge-storage kinetics and poor long-term stability. The results reported in this study demonstrate that charge-storage processes directed at 2D morphologies of surfaces and few layers of sheets are an exciting direction for improving the energy and power density of electrode materials for NIBs.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
757349
Fluoroethylene carbonate, 99%
C3H3FO3