EMAIL THIS PAGE TO A FRIEND
Analytica chimica acta
Impact of storage conditions on the urinary metabolomics fingerprint.
PMID 27998490
Abstract
Urine stability during storage is essential in metabolomics to avoid misleading conclusions or erroneous interpretations. Facing the lack of comprehensive studies on urine metabolome stability, the present work performed a follow-up of potential modifications in urinary chemical profile using LC-HRMS on the basis of two parameters: the storage temperature (+4xa0°C,xa0-20xa0°C,xa0-80xa0°C and freeze-dried stored atxa0-80xa0°C) and the storage duration (5-144 days). Both HILIC and RP chromatographies have been implemented in order to globally monitor the urinary metabolome. Using an original data processing associated to univariate and multivariate data analysis, our study confirms that chemical profiles of urine samples stored atxa0+4xa0°C are very rapidly modified, as observed for instance for compounds such as:N-acetyl Glycine, Adenosine, 4-Amino benzoic acid, N-Amino diglycine, creatine, glucuronic acid, 3-hydroxy-benzoic acid, pyridoxal, l-pyroglutamic acid, shikimic acid, succinic acid, thymidine, trigonelline and valeryl-carnitine, while it also demonstrates that urine samples stored atxa0-20xa0°C exhibit a global stability over a long period with no major modifications compared toxa0-80xa0°C condition. This study is the first to investigate long term stability of urine samples and report potential modifications in the urinary metabolome, using both targeted approach monitoring individually a large number (nxa0>xa0200) of urinary metabolites and an untargeted strategy enabling assessing for global impact of storage conditions.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
MSCAL5 ProteoMass LTQ/FT-Hybrid ESI Pos. Mode Cal Mix