EMAIL THIS PAGE TO A FRIEND
Oncology reports
miR-196b/miR-1290 participate in the antitumor effect of resveratrol via regulation of IGFBP3 expression in acute lymphoblastic leukemia.
PMID 28000876
Abstract
MicroRNAs play critical roles in the progression of acute lymphoblastic leukemia (ALL). Previous studies have indicated that miR-196b and miR-1290 play critical roles in T-cell ALL (T-ALL) and B-cell ALL (B-ALL), respectively. Resveratrol, a natural edible polyphenolic phytoalexin, possesses certain anticancer activities. Nevertheless, the mechanism involved in the regulation of ALL by resveratrol is still poorly understood. The present study aimed to reveal the potential mechanism underlying the antitumor effect of resveratrol in ALL focusing on miRNAs. Research indicates that insulin-like growth factor binding proteinxa03 (IGFBP3) plays a critical role in the aetiology of ALL. In the present study, we first demonstrated that the expression of IGFBP3 was decreased in ALL patients. We further identified that miR-196b and miR-1290 were overexpressed in T-ALL TALL-104 and B-ALL SUP-B15 cell lines, respectively. Moreover, resveratrol markedly decreased the overexpression of miR-196b/miR-1290 and elevated IGFBP3 expression in the ALL cell lines. As an miR-196b/miR-1290 inhibitor, resveratrol was further demonstrated to exert antitumor effects on ALL cells including antiproliferation, cell cycle arrest, apoptosis and inhibition of migration. Dual-luciferase reporter assay revealed that miR-196b/miR-1290 directly bound to the 3'-untranslated (3'-UTR) region of IGFBP3 mRNA. Moreover, we observed that IGFBP3 short interfering RNA reversed the antitumor activity of resveratrol against ALL cells. Taken together, the present study provides evidence that resveratrol targets miR-196b and miR-1290 for its antitumor activity in T-ALL and B-ALL, respectively. The present study also confirms that both miR‑196b and miR-1290 target the IGFBP3 3'-UTR and are potential therapeutic targets for ALL.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
EHU132051 MISSION® esiRNA, esiRNA human IGFBP3 (esiRNA1)