EMAIL THIS PAGE TO A FRIEND
Journal of clinical microbiology
Evaluation of Pyrrolidonyl Arylamidase Activity in Staphylococcus delphini.
PMID 28003425
Abstract
Clinical reference textbooks lack data for pyrrolidonyl arylamidase (PYR) activity in Staphylococcus delphini This study evaluated PYR activities of 21 S. delphini strains by reference broth, rapid disc, and rapid slide methods. Species and subgroup identifications were confirmed by nucleic acid-based methods and included nine group A and 12 group B strains. Testing by rapid PYR methods with products from four manufacturers was performed at two testing locations, and, with the exception of one strain tested at one location using reagents from one manufacturer, each S. delphini strain tested positive for PYR activity. Therefore, PYR may be a useful single-test adjunct for distinguishing Staphylococcus aureus from S. delphini and other members of the Staphylococcus intermedius group.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
P5891
L-Pyroglutamic acid 2-naphthylamide, ≥99% (TLC)
C15H14N2O2