EMAIL THIS PAGE TO A FRIEND
Human molecular genetics
Cellular, biochemical and molecular changes in muscles from patients with X-linked myotubular myopathy due to MTM1 mutations.
PMID 28007904
Abstract
Centronuclear myopathies are early-onset muscle diseases caused by mutations in several genes including MTM1, DNM2, BIN1, RYR1 and TTN. The most severe and often fatal X-linked form of myotubular myopathy (XLMTM) is caused by mutations in the gene encoding the ubiquitous lipid phosphatase myotubularin, an enzyme specifically dephosphorylating phosphatidylinositol-3-phosphate and phosphatidylinositol-3,5-bisphosphate. Because XLMTM patients have a predominantly muscle-specific phenotype a number of pathogenic mechanisms have been proposed, including a direct effect of the accumulated lipid on the skeletal muscle calcium channel ryanodine receptor 1, a negative effect on the structure of intracellular organelles and defective autophagy. Animal models knocked out for MTM1 show severe reduction of ryanodine receptor 1 mediated calcium release but, since knocking out genes in animal models does not necessarily replicate the human phenotype, we considered it important to study directly the effect of MTM1 mutations on patient muscle cells. The results of the present study show that at the level of myotubes MTM1 mutations do not dramatically affect calcium homeostasis and calcium release mediated through the ryanodine receptor 1, though they do affect myotube size and nuclear content. On the other hand, mature muscles such as those obtained from patient muscle biopsies exhibit a significant decrease in expression of the ryanodine receptor 1, a decrease in muscle-specific microRNAs and a considerable up-regulation of histone deacetylase-4. We hypothesize that the latter events consequent to the primary genetic mutation, are the cause of the severe decrease in muscle strength that characterizes these patients.