EMAIL THIS PAGE TO A FRIEND
Analytica chimica acta
Ultra-sensitive HPLC-photochemical reaction-luminol chemiluminescence method for the measurement of secondary amines after nitrosation.
PMID 28010842
Abstract
A novel method for the determination of secondary amines at the nanomolar level was developed. The method is based on the nitrosation reaction of secondary amines, with the generated N-nitrosamines being measured using an HPLC separation, photochemical reaction, and chemiluminescence detection system. The efficient nitrosation of secondary amines was performed using sodium nitrite (200xa0mM) and acetic acid (0.8xa0M) at 80xa0°C over 60xa0min. Although compounds bearing OH and SH functional groups also underwent the nitrosation reaction, the sensitivity of these compounds was 1000 times lower than that of the secondary amines. Our method was applied to the determination of low molecular weight secondary amines, including dimethylamine, morpholine, pyrrolidine, diethylamine, and piperidine, giving method detection limits of 0.7xa0nM, 0.2xa0nM, 0.4xa0nM, 0.7xa0nM, and 1.5xa0nM, respectively. The calibration curves were linear in the range of 5-100xa0nM. We then applied this method for the detection and quantification of these secondary amines in samples of tap water, river water, treated wastewater, and sea water. Dimethylamine was detected at concentrations up to 15.4xa0nM, <0.7xa0nM, and 48.5xa0nM in tap water, river water, and treated wastewater samples, respectively, with recoveries ranging from 94 to 103%. Other amines were also detected at nanomolar levels. These results indicate that our proposed method can be applied to the analysis of secondary amines in various environmental water samples. To the best of our knowledge, the proposed method is one of the most sensitive and selective methods for the determination of secondary amines without pre-concentration steps.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
A4562
5-(N,N-Dimethyl)amiloride hydrochloride
C8H12ClN7O · HCl
P5891
L-Pyroglutamic acid 2-naphthylamide, ≥99% (TLC)
C15H14N2O2