EMAIL THIS PAGE TO A FRIEND
Journal of medical entomology
Toxicity and Synergistic Activities of Chalcones Against Aedes aegypti (Diptera: Culicidae) and Drosophila melanogaster (Diptera: Drosophilidae).
PMID 28011724
Abstract
Mosquito-borne illnesses are of great concern throughout the world, and chemical insecticides are commonly employed to decrease mosquito populations. However, the developmental insecticide pipeline for vector control has primarily been filled by repurposed agricultural products, and is hampered by their widespread use and insecticide resistance. The present study was performed in the search for new chemical insecticides or insecticide synergists. Screening of 31 chalcone analogs was performed using Aedes aegypti (Linnaeus) first-instar larval toxicity assay, and oral feeding to Drosophila melanogaster's proper authority should be (Meigen). Synergism studies were performed by topically applying chalcones to adult female Ae. aegypti mosquitoes to examine its impact on activity of carbaryl, which was compared to piperonyl butoxide alone. Fourteen chalcone analogs had LC50 values in the range of 0.4-38 ppm against first-instar Ae. aegypti larvae, and three chalcones displayed toxicity against D. melanogaster via feeding (LC50 values ranged from 146-214 μg/ml). Two chalcones synergized carbaryl toxicity against adult Ae. aegypti with efficacy similar to piperonyl butoxide. As a result, it is concluded that chalcones may serve as novel insecticides and synergists after further structural optimization.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
68783
Licochalcone A, ≥96.0% (HPLC)
C21H22O4
SML0505
Tienilic Acid, ≥98% (HPLC)
C13H8Cl2O4S