EMAIL THIS PAGE TO A FRIEND
Journal of physiology and pharmacology : an official journal of the Polish Physiological Society
Bioavailability of aspirin in rats comparing the drug's uptake into gastrointestinal tissue and vascular and lymphatic systems: implications on aspirin's chemopreventive action.
PMID 28011944
Abstract
Aspirin is an effective analgesic and antiplatelet drug that in addition to its ability to reduce pain, inflammation and fever, appears to have efficacy in the prevention/treatment of a range of diseases including heart disease, numerous cancers and Alzheimer's. It is important to understand the bioavailability of aspirin and its major metabolite, salicylic acid, since dosage and route of administration can vary for treating differing diseases, and the major side-effects of aspirin, upper gastrointestinal ulceration and bleeding, are dose-dependent. We examined the time course for gastroduodenal uptake of aspirin and the appearance of its major metabolite salicylic acid in blood and lymph after intragastric (to simulate oral) and intraduodenal (to simulate enteric-coating) dosing in rats. Results show that after intragastric dosing, intact aspirin is absorbed primarily by the gastric mucosa and to a lesser extent by the duodenal mucosa. When aspirin is dosed intragastrically or intraduodenally, a much greater concentration of aspirin enters the lymph than the blood. In contrast, the concentration of salicylic acid was higher in blood than in lymph. Lymph levels of both aspirin and salicylic acid were sufficiently high so as to perform a pharmacologic function there, possibly as a chemopreventive agent against colon cancer and potentially the metastatic spread of non-gastrointestinal cancers.