EMAIL THIS PAGE TO A FRIEND
Food and chemical toxicology : an international journal published for the British Industrial Biological Research Association
The genotoxic potency of glycidol established from micronucleus frequency and hemoglobin adduct levels in mice.
PMID 28012894
Abstract
Glycidol is a genotoxic animal carcinogen that has raised concern due to its presence in food, as glycidyl fatty acid esters. Here we investigated the genotoxicity of glycidol in BalbC mice (0-120xa0mg/kg) by monitoring the induction of micronuclei in peripheral blood as a marker of chromosomal damage. The scoring of the micronuclei was assessed by flow cytometry. In the treated mice, the internal dose of glycidol, expressed as area under the concentration-time curve, AUC (molxa0×xa0L(-1)xa0×xa0h; Mh), was measured by dihydroxypropyl adducts to hemoglobin (Hb). The study showed that glycidol induced linear dose-dependent increases of Hb adducts (20xa0pmol/g Hb per mg/kg) and of micronuclei frequencies (12‰ per mMh). Compared to calculations based on administered dose, an improved dose-response relationship was observed when considering internal dose, achieved through the applied combination of sensitive techniques used for the scoring of micronuclei and AUC estimation of glycidol in the same mice. By comparing with earlier studies on micronuclei induction in mice exposed to ionizing radiation we estimated the radiation dose equivalent (rad-eq.) of glycidol to be ca 15xa0rad-eq./mMh.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
390062
Thiazole Orange, Dye content ~90 %
C26H24N2O3S2