EMAIL THIS PAGE TO A FRIEND
Microbial pathogenesis
The (95)(Δ)G mutation in the 5'untranslated region of the norA gene increases efflux activity in Staphylococcus epidermidis isolates.
PMID 28017900
Abstract
In the Staphylococcus aureus ATCC25923 strain, the flqB mutation in the 5'untranslated region (5'UTR) of the norA gene causes increased norA mRNA expression and high efflux activity (HEA). The involvement of the norA gene 5'UTR in HEA has not been explored in S.xa0epidermidis; therefore, we examined the function of this region in S.xa0epidermidis clinical isolates. The selection of isolates with HEA was performed based on ethidium bromide (EtBr) MIC values and efflux efficiency (EF) using the semi-automated fluorometric method. The function of the 5'UTR was studied by quantifying the levels of norA expression (RT-qPCR) and by identifying 5'UTR mutations by sequence analysis. Only 10 isolates from a total of 165 (6.1%) had HEA (EtBr MICxa0=xa0300xa0μg/ml and EF ranged from 48.4 to 97.2%). Eight of 10 isolates with HEA had the 5'UTR (95)(Δ)G mutation. Isolates carrying the (95)(Δ)G mutation had higher levels of norA expression compared with those that did not. To corroborate that the (95)(Δ)G mutation is involved in HEA, a strain adapted to EtBr was obtained inxa0vitro. This strain also presented the (95)(Δ)G mutation and had a high level of norA expression and EF, indicating that the (95)(Δ)G mutation is important for the HEA phenotype. The (95)(Δ)G mutation produces a different structure in the Shine-Dalgarno region, which may promote better translation of norA mRNA. To our knowledge, this is the first report to demonstrate the participation of the 5'UTR (95)(Δ)G mutation of the norA gene in the HEA phenotype of S.xa0epidermidis isolates. Here, we propose that the efflux of EtBr is caused by an increment in the transcription and/or translation of the norA gene.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
C2759
Carbonyl cyanide 3-chlorophenylhydrazone, ≥97% (TLC), powder
C9H5ClN4