EMAIL THIS PAGE TO A FRIEND
Nanoscale research letters
TiO2 Nanotube-Carbon (TNT-C) as Support for Pt-based Catalyst for High Methanol Oxidation Reaction in Direct Methanol Fuel Cell.
PMID 28032325
Abstract
In this study, TiO2 nanotubes (TNTs) were synthesized via a hydrothermal method using highly concentrated NaOH solutions varying from 6 to 12xa0M at 180xa0°C for 48xa0h. The effects of the NaOH concentration and the TNT crystal structure on the performance for methanol oxidation were investigated to determine the best catalyst support for Pt-based catalysts. The results showed that TNTs produced with 10xa0M NaOH exhibited a length and a diameter of 550 and 70xa0nm, respectively; these TNTs showed the best nanotube structure and were further used as catalyst supports for a Pt-based catalyst in a direct methanol fuel cell. The synthesized TNT and Pt-based catalysts were analysed by FESEM, TEM, BET, EDX, XRD and FTIR. The electrochemical performance of the catalysts was investigated using cyclic voltammetry (CV) and chronoamperometric (CA) analysis to further understand the methanol oxidation in the direct methanol fuel cell (DMFC). Finally, the result proves that Pt-Ru/TNT-C catalyst shows high performance in methanol oxidation as the highest current density achieved at 3.3xa0mA/cm(2) (normalised by electrochemically active surface area) and high catalyst tolerance towards poisoning species was established.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
208523
Ruthenium(III) chloride, Ru content 45-55%
Cl3Ru