EMAIL THIS PAGE TO A FRIEND
Orthopaedic surgery
N-acetylcysteine Attenuates Cobalt Nanoparticle-Induced Cytotoxic Effects through Inhibition of Cell Death, Reactive Oxygen Species-related Signaling and Cytokines Expression.
PMID 28032714
Abstract
Complex cobalt-chromium alloys, bearing surfaces of the second-generation metal-on-metal (MoM) hip prostheses, are subject to wear and generate cobalt nanoparticles (CoNPs). CoNPs could reduce cellular viability, activate the mitogen-activated protein kinase (MAPK) pathway and increase cell apoptosis via reactive oxygen species (ROS). However, the detailed mechanisms of ROS functioning on CoNP-mediated signaling molecules and cytotoxicity has not yet been fully demonstrated. The present study investigated the functional role of N-acetylcysteine (NAC) in reversing the activation of ROS signaling pathways triggered by CoNPs in normal mice kidney cells (TCMK-1 cells). After being pretreated with NAC, TCMK-1 cells were treated with 300-700 μmol/L CoNPs, then, CCK-8 assay was used to verify the survival of TCMK-1 cells. Annexin V/PI staining was performed to investigate the apoptosis of TCMK-1 cells after NAC and different concentrations of CoNP treatments. In addition, western blot was performed to identify the cytokine (p-ERK, p-p38, and p-JNK) expression of the ROS-related MAPK signaling pathway. Apoptosis rate of TCMK-1 cells was increased obviously after different concentrations of CoNP treatment. However, TCMK-1 cells, pretreated with NAC, exhibited a significantly decreased apoptosis rate. In addition, p-ERK, p-p38, and p-JNK expressions were increased with CoNP treatment, which indicated that CoNPs could activate the MAPK pathway. Interestingly, this entire stimulated phenomenon by CoNPs was reversed with NAC treatment. These findings indicated that NAC could reverse CoNP-induced cytotoxicity by inhibiting ROS-induced cell death and cytokine expression. To our knowledge, this is the first report that describes how CoNP-induced cytotoxicity in TCMK-1 cells could be attenuated by anti-oxidative agents (NAC), which may function through inhibition of cell death and ROS.