EMAIL THIS PAGE TO A FRIEND
Neuroscience letters
Increased susceptibility to Aβ toxicity in neuronal cultures derived from familial Alzheimer's disease (PSEN1-A246E) induced pluripotent stem cells.
PMID 28034781
Abstract
Alzheimer's disease (AD) is the most common cause of late-life dementia and represents one of the leading causes of death worldwide. The generation of induced pluripotent stem cells (iPSC) has facilitated the production and differentiation of stem cells from patients somatic cells, offering new opportunities to model AD and other diseases in vitro. In this study, we generated iPSCs from skin fibroblasts obtained from a healthy individual, as well as sporadic (sAD) and familial AD (fAD, PSEN1-A246E mutation) patients. iPSC lines were differentiated into neuronal precursors (iPSC-NPCs) and neurons that were subjected to amyloid beta (Aβ) toxicity assays. We found that neurons derived from the fAD patient have a higher susceptibility to Aβ1-42 oligomers compared with neurons coming from healthy and sAD individuals. Our findings suggest that neurons from patients with PSEN1-A246E mutation have intrinsic properties that make them more susceptible to the toxic effects of Aβ1-42 oligomers in the AD brain.