EMAIL THIS PAGE TO A FRIEND
Free radical biology & medicine
Halofuginone enhances the chemo-sensitivity of cancer cells by suppressing NRF2 accumulation.
PMID 28039084
Abstract
The KEAP1-NRF2 system regulates the cellular defence against oxidative and xenobiotic stresses. NRF2 is a transcription factor that activates the expression of cytoprotective genes encoding antioxidative, detoxifying and metabolic enzymes as well as transporters. Under normal conditions, KEAP1 represses NRF2 activity by degrading the NRF2 protein. When cells are exposed to stresses, KEAP1 stops promoting NRF2 degradation, and NRF2 rapidly accumulates and activates the transcription of target genes. Constitutive accumulation of NRF2 via a variety of mechanisms that disrupt KEAP1-mediated NRF2 degradation has been observed in various cancer types. Constitutive NRF2 accumulation confers cancer cells with a proliferative advantage as well as resistance to anti-cancer drugs and radiotherapies. To suppress the chemo- and radio-resistance of cancer cells caused by NRF2 accumulation, we conducted high-throughput chemical library screening for NRF2 inhibitors and identified febrifugine derivatives. We found that application of the less-toxic derivative halofuginone in a low dose range rapidly reduced NRF2 protein levels. Halofuginone induced a cellular amino acid starvation response that repressed global protein synthesis and rapidly depleted NRF2. Halofuginone treatment ameliorated the resistance of NRF2-addicted cancer cells to anti-cancer drugs both in vitro and in vivo. These results provide preclinical proof-of-concept evidence for halofuginone as an NRF2 inhibitor applicable to treatment of chemo- and radio-resistant forms of cancer.