EMAIL THIS PAGE TO A FRIEND
Food microbiology
Cloning the putative gene of vinyl phenol reductase of Dekkera bruxellensis in Saccharomyces cerevisiae.
PMID 28040186
Abstract
Vinylphenol reductase of Dekkera bruxellensis, the characteristic enzyme liable for "Brett" sensory modification of wine, has been recently recognized to belong to the short chain dehydrogenases/reductases family. Indeed, a preliminary biochemical characterisation has conferred to the purified protein a dual significance acting as superoxide dismutase and as a NADH-dependent reductase. The present study aimed for providing a certain identification of the enzyme by cloning the VPR gene in S.xa0cerevisiae, a species not producing ethyl phenols. Transformed clones of S.xa0cerevisiae resulted capable of expressing a biologically active form of the heterologous protein, proving its role in the conversion of 4-vinyl guaiacol to 4-ethyl guaiacol. A VPR specific protein activity of 9xa0±xa00.6xa0mU/mg was found in crude extracts of S.xa0cerevisiae recombinant strain. This result was confirmed in activity trials carried out with the protein purified from transformant cells of S.xa0cerevisiae by a his-tag purification approach; in particular, VPR-enriched fractions showed a specific activity of 1.83xa0±xa00.03xa0U/mg at pH 6.0. Furthermore, in agreement with literature, the purified protein behaves like a SOD, with a calculated specific activity of approximatively 3.41xa0U/mg. The comparative genetic analysis of the partial VPR gene sequences from 17 different D.xa0bruxellesis strains suggested that the observed polymorphism (2.3%) and the allelic heterozygosity state of the gene do not justify the well described strain-dependent character in producing volatile phenols of this species. Actually, no correlation exists between genotype membership of the analysed strains and their capability to release off-flavours. This work adds valuable knowledge to the study of D.xa0bruxellensis wine spoilage and prepare the ground for interesting future industrial applications.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
P6611
HIS-Select® Nickel Affinity Gel