EMAIL THIS PAGE TO A FRIEND
Clinical and experimental immunology
Programmed death-1 ligands 1 and 2 expression in cutaneous squamous cell carcinoma and their relationship with tumour- infiltrating dendritic cells.
PMID 28052400
Abstract
The programmed death-1 (PD-1) receptor ligands, PD-L1 and PD-L2, are co-stimulatory molecules that contribute to the negative regulation of T lymphocyte activation. It is still unclear whether there is correlation between PD-L1 or PD-L2 and tumour-infiltrating dendritic cells (TIDCs) in cutaneous squamous cell carcinoma (CSCC). The aim of this study was to analyse PD-L1 and PD-L2 expression and dendritic cells infiltration in tumour tissue of CSCC patients and investigate their clinical significance. Immunohistochemical analysis was used to evaluate the expression of PD-L1, PD-L2, CD1a and CD83 in 61 CSCC tissues. The immunofluoresence double-labelling technique was performed to detect the co-expression of PD-L1 or PD-L2 and CD1a or CD83 in tumour tissues. We found that 25 of 61 cases CSCC (40·98%) exhibited positivity for PD-L1, whereas 37 of 61 cases CSCC (60·66%) exhibited positivity for PD-L2. A higher percentage of CD1a-positive cases were observed on both PD-L1-positive and PD-L2-positive specimens compared with that of CD83-positive cases (92·29% versus 37·60%, 83·20% versus 33·16%). The expression of PD-L1 and PD-L2 on CD1a(+) cells was significantly higher than that on CD83(+) cells in tumour tissues of CSCC patients. Furthermore, the expression rate of PD-L1 was associated with UICC stage, and the expression rate of PD-L2 was associated with predominant differentiation and tumour size in CSCC. Our results indicated that higher expression of PD-L1 and PD-L2 on CD1a(+) cells than that on CD83(+) cells in CSCC tumour tissues may contribute to negative regulation in anti-tumour immune responses.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
P26600
2-Phenylindole, technical grade, 95%
C14H11N