EMAIL THIS PAGE TO A FRIEND
Cell death & disease
Early nucleolar disorganization in Dictyostelium cell death.
PMID 28055008
Abstract
Cell death occurs in all eukaryotes, but it is still not known whether some core steps of the cell death process are conserved. We investigated this using the protist Dictyostelium. The dissection of events in Dictyostelium vacuolar developmental cell death was facilitated by the sequential requirement for two distinct exogenous signals. An initial exogenous signal (starvation and cAMP) recruited some cells into clumps. Only within these clumps did subsequent cell death events take place. Contrary to our expectations, already this initial signal provoked nucleolar disorganization and irreversible inhibition of rRNA and DNA synthesis, reflecting marked cell dysfunction. The initial signal also primed clumped cells to respond to a second exogenous signal (differentiation-inducing factor-1 or c-di-GMP), which led to vacuolization and synthesis of cellulose encasings. Thus, the latter prominent hallmarks of developmental cell death were induced separately from initial cell dysfunction. We propose that (1) in Dictyostelium vacuolization and cellulose encasings are late, organism-specific, hallmarks, and (2) on the basis of our observations in this protist and of similar previous observations in some cases of mammalian cell death, early inhibition of rRNA synthesis and nucleolar disorganization may be conserved in some eukaryotes to usher in developmental cell death.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
A9414
Agarose, low gelling temperature, BioReagent, for molecular biology