EMAIL THIS PAGE TO A FRIEND
The Journal of biological chemistry
The Receptor-interacting Serine/Threonine Protein Kinase 1 (RIPK1) Regulates Progranulin Levels.
PMID 28069809
Abstract
Progranulin (PGRN), a secreted growth factor, is a key regulator of inflammation and is genetically linked to two common and devastating neurodegenerative diseases. Haploinsufficiency mutations in GRN, the gene encoding PGRN, cause frontotemporal dementia (FTD), and a GRN SNP confers significantly increased risk for Alzheimer's disease (AD). Because cellular and animal data indicate that increasing PGRN can reverse phenotypes of both FTD and AD, modulating PGRN level has been proposed as a therapeutic strategy for both diseases. However, little is known about the regulation of PGRN levels. In this study, we performed an siRNA-based screen of the kinome to identify genetic regulators of PGRN levels in a rodent cell-based model system. We found that knocking down receptor-interacting serine/threonine protein kinase 1 (Ripk1) increased both intracellular and extracellular PGRN protein levels by increasing the translation rate of PGRN without affecting mRNA levels. We observed this effect in Neuro2a cells, wild-type primary mouse neurons, and Grn-haploinsufficient primary neurons from an FTD mouse model. We found that the effect of RIPK1 on PGRN is independent of the kinase activity of RIPK1 and occurs through a novel signaling pathway. These data suggest that targeting RIPK1 may be a therapeutic strategy in both AD and FTD.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
861677
DL-Cysteine, technical grade
C3H7NO2S