EMAIL THIS PAGE TO A FRIEND
Journal of translational medicine
Antibiotics-induced depletion of mice microbiota induces changes in host serotonin biosynthesis and intestinal motility.
PMID 28086815
Abstract
The gastrointestinal motility is affected by gut microbiota and the relationship between them has become a hot topic. However, mechanisms of microbiota in regulating motility have not been well defined. We thus investigated the effect of microbiota depletion by antibiotics on gastrointestinal motility, colonic serotonin levels, and bile acids metabolism. After 4xa0weeks with antibiotics treatments, gastrointestinal and colon transit, defecation frequency, water content, and other fecal parameters were measured and analyzed in both wild-type and antibiotics-treated mice, respectively. Contractility of smooth muscle, serotonin levels, and bile acids levels in wild-type and antibiotics-treated mice were also analyzed. After antibiotics treatment, the richness and diversity of intestinal microbiota decreased significantly, and the fecal of mice had less output (Pxa0
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
M0262
Methyl cellulose, viscosity: 400 cP