EMAIL THIS PAGE TO A FRIEND
BMC neuroscience
Poly(ADP-ribose) polymerase inhibitors activate the p53 signaling pathway in neural stem/progenitor cells.
PMID 28095779
Abstract
Poly(ADP-ribose) polymerase 1 (PARP-1), which catalyzes poly(ADP-ribosyl)ation of proteins by using NAD(+) as a substrate, plays a key role in several nuclear events, including DNA repair, replication, and transcription. Recently, PARP-1 was reported to participate in the somatic cell reprogramming process. Previously, we revealed a role for PARP-1 in the induction of neural apoptosis in a cellular model of cerebral ischemia and suggested the possible use of PARP inhibitors as a new therapeutic intervention. In the present study, we examined the effects of PARP inhibitors on neural stem/progenitor cells (NSPCs) of the mouse brain. PARP-1 was more abundant and demonstrated higher activity in NSPCs than in mouse embryonic fibroblasts. Treatment with PARP inhibitors suppressed the formation of neurospheres by NSPCs through the suppression of cell cycle progression and the induction of apoptosis. In order to identify the genes responsible for these effects, we investigated gene expression profiles by microarray analyses and found that several genes in the p53 signaling pathway were upregulated, including Cdkn1a, which is critical for cell cycle control, and Fas, Pidd, Pmaip1, and Bbc3, which are principal factors in the apoptosis pathway. Inhibition of poly(ADP-ribosyl)ation increased the levels of p53 protein, but not p53 mRNA, and enhanced the phosphorylation of p53 at Ser18. Experiments with specific inhibitors and also shRNA demonstrated that PARP-1, but not PARP-2, has a role in the regulation of p53. The effects of PARP inhibitors on NSPCs were not observed in Trp53 (-/-) NSPCs, suggesting a key role for p53 in these events. On the basis of the finding that PARP inhibitors facilitated the p53 signaling pathway, we propose that poly(ADP-ribosyl)ation contributes to the proliferation and self-renewal of NSPCs through the suppression of p53 activation.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
I138
1,5-Isoquinolinediol, ≥98% (HPLC), powder
C9H7NO2