EMAIL THIS PAGE TO A FRIEND
Vox sanguinis
Non-polar lipids accumulate during storage of transfusion products and do not contribute to the onset of transfusion-related acute lung injury.
PMID 28097705
Abstract
The accumulation of non-polar lipids arachidonic acid, 5-hydroxyeicosatetraenoic acid (HETE), 12-HETE and 15-HETE during storage of transfusion products may play a role in the onset of transfusion-related acute lung injury (TRALI), a syndrome of respiratory distress after transfusion. We investigated non-polar lipid accumulation in red blood cells (RBCs) stored for 42 days, plasma stored for 7 days at either 4 or 20°C and platelet (PLT) transfusion products stored for 7 days. Furthermore, we investigated whether transfusion of RBCs with increased levels of non-polar lipids induces TRALI in a 'two-hit' human volunteer model. All products were produced following Dutch Blood Bank protocols and are according to European standards. Non-polar lipids were measured with high-performance liquid chromotography followed by mass spectrometry. All non-polar lipids increased in RBCs after 21 days of storage compared to baseline. The non-polar lipid concentration in plasma increased significantly, and the increase was even more pronounced in products stored at 20°C. In platelets, baseline levels of 5-HETE and 15-HETE were higher than in RBCs or plasma. However, the non-polar lipids did not change significantly during storage of PLT products. Infusion of RBCs with increased levels of non-polar lipids did not induce TRALI in LPS-primed human volunteers. We conclude that non-polar lipids accumulate in RBC and plasma transfusion products and that accumulation is temperature dependent. Accumulation of non-polar lipids does not appear to explain the onset of TRALI (Dutch Trial Register - NTR4455).
Related Materials