PloS one
An Approach to Elucidate NBS1 Function in DNA Repair Using Frequent Nonsynonymous Polymorphism in Wild Medaka (Oryzias latipes) Populations.
PMID 28107384
Abstract
Nbs1 is one of the genes responsible for Nijmegen breakage syndrome, which is marked with high radiosensitivity. In human NBS1 (hNBS1), Q185E polymorphism is known as the factor to cancer risks, although its DSB repair defect has not been addressed. Here we investigated the genetic variations in medaka (Oryzias latipes) wild populations, and found 40 nonsynonymous single nucleotide polymorphisms (SNPs) in medaka nbs1 (olnbs1) gene within 5 inbred strains. A mutation to histidine in Q170 residue in olNbs1, which corresponds to Q185 residue of hNBS1, was widely distributed in the closed colonies derived from the eastern Korean population of medaka. Overexpression of H170 type olNbs1 in medaka cultured cell lines resulted in the increased accumulation of olNbs1 at laser-induced DSB sites. Autophosphorylation of DNA-dependent protein kinase at T2609 was suppressed after the γ-ray irradiation, which was followed by prolonged formation of γ-H2AX foci and delayed DSB repair. These findings suggested that the nonsynonymous SNP (Q170H) in olnbs1, which induced DSB repair defects, is specifically distributed in the eastern Korean population of medaka. Furthermore, examination using the variation within wild populations might provide a novel method to characterize a driving force to spread the disease risk alleles.
Related Materials