EMAIL THIS PAGE TO A FRIEND
Talanta
Use of quantum dot beads-labeled monoclonal antibody to improve the sensitivity of a quantitative and simultaneous immunochromatographic assay for neuron specific enolase and carcinoembryonic antigen.
PMID 28107959
Abstract
Detection of multiplex tumor markers was of great importance for cancer diagnosis. Immunochromatographic test strip (ICTS) was the most frequently-used point-of-care detection means. Herein, a convenient and fast method for simultaneous quantitative detection of neuron specific enolase (NSE) and carcinoembryonic antigen (CEA) was developed based on ICTS using quantum dot beads (QBs) as marking material. Good monodispersity, high colloidal stability and carboxyl-modified (COOH-) QBs were used. For this method, two test lines were applied to the NC membrane for simultaneous analysis of CEA and NSE respectively. The ideal limit of CEA and NSE detection was 0.0378ng/mL and 0.0426ng/mL with scarcely any cross-reactivity. Moreover, the fluorescent signal intensity of the nitrocellulose membrane could be easily read out in the cooperation of the "Handing" system without professional operators. The possible clinical utilization of this platform was demonstrated by detecting 100 clinic human serums. The result showed that the platform had sensitivity of 99% and 97% for CEA and NSE, while the specificity was 97% and 100% respectively. Our results indicated that the QBs based ICTS not only owning the ability of sensitive and specific simultaneous detection of CEA and NSE, but also showing the potential in developing this ICTS into a routine part of early lung cancer diagnosis.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
729817
2-Chloroethyl acrylate, contains >100 ppm MEHQ as inhibitor, 97%
C5H7ClO2