EMAIL THIS PAGE TO A FRIEND
Biomaterials
Inxa0vivo near-infrared imaging and phototherapy of tumors using a cathepsin B-activated fluorescent probe.
PMID 28113107
Abstract
The development of multifunctional reagents for simultaneous specific near-infrared (NIR) imaging and phototherapy of tumors is of great significance. This work describes the design of a cathepsin B-activated fluorescent probe (CyA-P-CyB) and its applications as an NIR imaging probe for tumor cells and as a phototherapy reagent for tumors. Inxa0vitro experiments demonstrated that CyA-P-CyB was activated via the cleavage of a peptide linker by cathepsin B in tumor cells to produce fluorescence in the NIR region based on a FRET mechanism. MTT assays showed that the phototoxicity of CyA-P-CyB toward cells depended on the activity of cathepsin B, and the probe exhibited specific phototoxicity toward tumor cells. CyA-P-CyB was also successfully applied to the inxa0vivo imaging and phototherapy of tumors. Histological analysis indicated that CyA-P-CyB had no cytotoxic effects on seven mouse tissues (lung, liver, heart, kidney, pancreas, spleen and brain) after the CyA-P-CyB treatment and laser irradiation.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
762016
3-Azido-1-propanamine, 90%
C3H8N4