EMAIL THIS PAGE TO A FRIEND
Oncology letters
Comparative proteomic analysis of paclitaxel resistance-related proteins in human breast cancer cell lines.
PMID 28123557
Abstract
Paclitaxel is widely used to treat various cancers; however, resistance to this drug is a major obstacle to breast cancer chemotherapy. To identify the proteins involved in paclitaxel resistance, the present study compared the proteomes of MCF-7 human breast cancer cells and its paclitaxel-resistant subclone MCF-7/PTX. Using two-dimensional gel electrophoresis and matrix-assisted laser desorption/ionization time of flight mass spectrometry, 11 upregulated and 12 downregulated proteins were identified in MCF-7/PTX cells compared with the parental cell line. These 23 proteins were functionally classified as stress-induced chaperones, metabolic enzymes and cytoskeletal proteins. The anti-apoptotic proteins, stress-70 protein, 78-kD glucose-regulated protein, peptidyl-prolyl cis-trans isomerase A (PPIA) and heterogeneous nuclear ribonucleoprotein H3, were also upregulated in MCF-7/PTX cells. Notably, knockdown of the stress-response chaperone PPIA using small interfering RNA in MCF-7/PTX cells restored their sensitivity to paclitaxel. These findings indicated that PPIA may have an important role in paclitaxel resistance in MCF-7/PTX cells.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
EHU107101 MISSION® esiRNA, esiRNA human PPIA (esiRNA1)