EMAIL THIS PAGE TO A FRIEND
Journal of hazardous materials
Nanostructured CoP: An efficient catalyst for degradation of organic pollutants by activating peroxymonosulfate.
PMID 28126574
Abstract
A new catalyst system of CoP/peroxymonosulfate (PMS) is presented, which achieved significant improvement in catalytic activity. Nanostructured CoP, obtained by a simple solid-state reaction, exhibited dramatic catalytic activity with 97.2% degradation of orange II of 100ppm within 4min. Moreover, the high efficiency could be reached for other phenolic pollutants, i.e., phenol and 4-chlorophenol. The reaction rate is much higher than the most reported catalysts. Effect of parameters on catalytic activity of the catalyst was studied in detail. Notably, initial pH of the solution had a slight negative effect on the catalytic performance over the pH range 4.07-10.92, suggesting that CoP has the great adaptability of pH. CoP/PMS demonstrated excellent anti-interference performance toward anions (Cl(-), NO3(-), and HCO3(-)). In addition, the pathway of degradation of orange II is proposed by analyzing its intermediates. Based on the XPS spectra of CoP, the identification of the reactive species (OH and SO4(-)) by electron paramagnetic resonance (EPR) analysis and quenching tests, a possible mechanism for activation of PMS by CoP was proposed. Considering the dramatic catalytic activity, a wide range of pH catalyst suited, CoP is believed to provide robust support for the promising industrial application of AOPs.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
228036
OXONE®, monopersulfate compound
HKO5S · 0.5HKO4S · 0.5K2O4S
P9625
Phenazine methosulfate, ≥90% (UV)
C14H14N2O4S