EMAIL THIS PAGE TO A FRIEND
Neuroscience letters
The naturally occurring mutation Y197C does not affect the expression or signaling of the human histamine H3 receptor.
PMID 28126588
Abstract
There is evidence for genetic polymorphism within the human histamine H3 receptor (hH3R), and a Tyr to Cys exchange at position 197 (Y197C), located in the amino terminus of the fifth transmembrane domain, has been reported. In this work we compared the expression and the pharmacological and signaling properties of wild-type (hH3RWT) and mutant (hH3RY197C) receptors transiently expressed in CHO-K1 cells. The hH3RY197C cDNA was created by overlap extension PCR amplification. Receptor expression and affinity were assessed by N-α-[methyl-(3)H]-histamine binding to cell membranes and intact cells. Receptor function was evaluated by stimulation of [(35)S]-GTPγS binding to cell membranes and by inhibition of forskolin-induced cAMP accumulation in intact cells. The hH3RWT and hH3RY197C were expressed at similar levels (761±68 and 663±66fmol/mg protein for membranes, and 13,434±1533 and 15,894±1884 receptors per cell, respectively). There were no significant differences in the affinities for H3R agonists or antagonists/inverse agonists between the hH3RWT and hH3RY197C, and the H3R agonist RAMH was similarly efficacious and potent to stimulate [(35)S]-GTPγS binding and to inhibit forskolin-induced cAMP accumulation. These results indicate that the Y197C mutation does not affect the expression, ligand affinity or signaling of the human H3 receptor.
Related Materials