EMAIL THIS PAGE TO A FRIEND
Scientific reports
Neutrophil gelatinase-associated lipocalin (NGAL) fails as an early predictor of contrast induced nephropathy in chronic kidney disease (ANTI-CI-AKI study).
PMID 28128223
Abstract
The aim of the study was to evaluate the diagnostic accuracy of urinary neutrophil gelatinase- associated lipocalin (uNGAL) in patients with chronic kidney disease (CKD) as an early biomarker for contrast induced acute kidney injury (CI-AKI) and to investigate whether patients with an uNGAL increase might benefit from an additional intravenous volume expansion with regard to CI-AKI-incidence. We performed a prospective randomized controlled trial in 617 CKD-patients undergoing intra-arterial angiography. Urinary NGAL was measured the day before and 4-6hrs after angiography. In the event of a significant rise of uNGAL patients were randomized either into Group A, who received intravenous saline post procedure or Group B, who did not receive post-procedural i.v. fluids. Ten patients (1.62%) exhibited a significant rise of uNGAL after angiography and were randomized of whom one developed a CI-AKI. In the entire cohort the incidence of CI-AKI was 9.4% (58 patients) resulting in a specificity of 98.4% (95% CI: 97.0-99.3%) and a sensitivity of 1.72% (95% CI: 0.044-9.2%) of uNGAL for the diagnosis of CI-AKI. In this study uNGAL failed to predict CI-AKI and was an inadequate triage tool to guide an early intervention strategy to prevent CI-AKI. URL: http://www.clinicaltrials.gov. Unique identifier: NCT01292317.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
469548
1,1,1-Trifluoro-5,5-dimethyl-2,4-hexanedione, 98%
C8H11F3O2