EMAIL THIS PAGE TO A FRIEND
Chemosphere
In vitro cytotoxic effects of DEHP-alternative plasticizers and their primary metabolites on a L929 cell line.
PMID 28129623
Abstract
Phthalic acid esters have been widely used to improve the plasticity of PVC medical devices. They carry a high exposure risk for both humans and the environment in clinical situations. Our study focuses on the cytotoxicity of alternative plasticizers. Postulated primary metabolites were synthesized, not being commercially available. Cytotoxicity assays were performed on L929 murine cells according to the ISO-EN 10993-5 standard design for the biocompatibility of medical devices. The tested concentrations of plasticizers (0.01, 0.05 and 0.1 mg/ml) covered the range likely to be found in biological fluids coming into direct contact with the medical devices. DEHP, DINP and DINCH were cytotoxic at the highest concentration (0.1 mg/ml) for 7 days of exposure. Their corresponding metabolites were found to be more cytotoxic, for the same concentration. By contrast, TOTM and its corresponding metabolite MOTM were not found to be cytotoxic. DEHA showed no cytotoxicity, but its corresponding monoester (MEHA) produced a cytotoxic effect at 0.05 mg/ml. In clinical situations, medical devices can release plasticizers, which can come into contact with patients. In vivo, the plasticizers are quickly transformed into primary metabolites. It is therefore important to measure the effects of both the plasticizers and their corresponding metabolites. Standard first-line cytotoxicity assays should be performed to ensure biocompatibility.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
92124
Tris(2-ethylhexyl) trimellitate, Selectophore
C33H54O6