EMAIL THIS PAGE TO A FRIEND
Soft matter
Patterning of perovskite-polymer films by wrinkling instabilities.
PMID 28138668
Abstract
Organic-inorganic perovskites are semiconductors used for applications in optoelectronics and photovoltaics. Micron and submicron perovskite patterns have been explored in semitransparent photovoltaic and lasing applications. In this work, we show that a polymeric medium can be used to create a patterned perovskite, by using a novel and inexpensive approach.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
806498
Methylammonium bromide
CH6BrN