EMAIL THIS PAGE TO A FRIEND
Global public health
The Taiwan Tobacco and Liquor Corporation: To 'join the ranks of global companies'.
PMID 28139964
Abstract
Until the late 1990s, the Taiwan Tobacco and Liquor Corporation (TTL) focused almost exclusively on serving the domestic market as a highly protected monopoly. This paper describes how the company has adopted a more outward looking strategy since 2000, with ambitions to become a regional, and eventually global, business by 2021. Drawing on company documents and industry sources, the paper argues that this shift in strategy was a direct reaction to the decline in domestic market share following liberalisation of the Taiwanese tobacco market and adoption of tougher domestic tobacco control measures. Market opening occurred as a result of pressure from the U.S. Trade Representative in the 1980s, as well as World Trade Organization membership in 2002. It is argued that TTL's efforts to globalise operations have been limited by bureaucratic company management and structures, and ongoing political tension between Taiwan and China. However, the relative success of TTL's alcohol branch, and potential détente as the Taiwanese government reaches out to improve relations with China, may provide TTL with new opportunities to achieve its goal of becoming a regional player with global ambitions. This article is part of the special issue 'The Emergence of Asian Tobacco Companies: Implications for Global Health Governance.'
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
12094
Docosanoic acid tryptamide, analytical standard
C32H54N2O