EMAIL THIS PAGE TO A FRIEND
Nature communications
β-arrestin-2 is an essential regulator of pancreatic β-cell function under physiological and pathophysiological conditions.
PMID 28145434
Abstract
β-arrestins are critical signalling molecules that regulate many fundamental physiological functions including the maintenance of euglycemia and peripheral insulin sensitivity. Here we show that inactivation of the β-arrestin-2 gene, barr2, in β-cells of adult mice greatly impairs insulin release and glucose tolerance in mice fed with a calorie-rich diet. Both glucose and KCl-induced insulin secretion and calcium responses were profoundly reduced in β-arrestin-2 (barr2) deficient β-cells. In human β-cells, barr2 knockdown abolished glucose-induced insulin secretion. We also show that the presence of barr2 is essential for proper CAMKII function in β-cells. Importantly, overexpression of barr2 in β-cells greatly ameliorates the metabolic deficits displayed by mice consuming a high-fat diet. Thus, our data identify barr2 as an important regulator of β-cell function, which may serve as a new target to improve β-cell function.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
P9767
Sodium palmitate, ≥98.5%
C16H31NaO2
V5754
Veratridine, ≥90% (HPLC), powder
C36H51NO11