EMAIL THIS PAGE TO A FRIEND
Saudi journal of biological sciences
In vitro study of biological activity of four strains of Burkholderia gladioli pv. agaricicola and identification of their bioactive metabolites using GC-MS.
PMID 28149165
Abstract
This research was carried out to study in vitro antibacterial activity of 4 strains of Burkholderia gladioli pv. agaricicola (Bga) against G+ve Bacillus megaterium and G-ve Escherichia coli, haemolytic activity against the cell membrane of erythrocytes, the production of extracellular hydrolytic enzymes and finally, the pathogenicity against Agaricus bisporus flesh blocks. Chemical structure of bioactive substances of the most bioactive strain (ICMP 11096) was established using gas chromatography-mass spectrometry (GC-MS). All the studied Bga strains inhibited the growth of the two tested bacteria although some growing substrates negatively influenced the antimicrobial substance production. The same Bga strains showed highly haemolytic activity and were able to produce 3 hydrolytic enzymes, i.e. chitinase, glucanase and protease. In pathogenicity assays, the considered Bga strains resulted virulent for A. bisporus. The GC-MS for compounds from Bga ICMP 11096 were compatible with the structure of two bioactive fatty acids identified as methyl stearate and ethanol 2-butoxy phosphate with mass spectrum m/e 298 and 398, respectively.