EMAIL THIS PAGE TO A FRIEND
Journal of thoracic disease
LMO4 is a prognostic marker involved in cell migration and invasion in non-small-cell lung cancer.
PMID 28149564
Abstract
The aims of this study were to analyze the association of LMO4 with non-small-cell lung cancer (NSCLC) survival rate, and to determine its functional role and signaling pathway in lung cancer. Immunohistochemistry (IHC) was used to detect the expression of LMO4 in NSCLC cell lines and tumor tissues. Migration and invasion ability was detected respectively by wound healing test and transwell test. Immunofluorescence and western blot were detected of AKT/PI3K pathway related genes MAPK, PI3K, AKT. LMO4 has high expression level of NSCLC cell lines and tumor tissues, and correlated with a lower survival rate. LMO4 can regulate the migration and invasion of NSCLC cells through the AKT/PI3K pathway. LMO4 could serve as a promising biomarker and therapeutic target for NSCLC.