EMAIL THIS PAGE TO A FRIEND
Journal of colloid and interface science
Synthesis and characterization of functionalized mesoprous SBA-15 decorated with Fe(3)O(4) nanoparticles for removal of Ce(III) ions from aqueous solution: ICP-OES detection and central composite design optimization.
PMID 28152439
Abstract
A selective adsorbent based on the modification of mesoprous SBA-15 with N,N'-bis(salicylidene)-1,3-ethylenediamine Schiff base and decorated with Fe3O4 nanoparticles (SBA-15-BSEA-Fe3O4-NPs) for Ce(III) ions removal was reported. The SBA-15-BSEA-Fe3O4-NPs was identified by XRD, FE-SEM, TEM, SEM, FT-IR, VSM, BET and BJH analysis. Central composite design (CCD) was applied to evaluate the main and interactive effects of adsorption variables and optimize the operational parameters. The important variable such as initial pH solution, SBA-15-BSEA-Fe3O4-NPs mass, shaking time and initial concentration of Ce(3+) ions were studied under batch mode. In desirability concession of 1.0 as optimum value for R% Ce(III), the level of factors was as follows: shaking time 80min, SBA-15-BSEA-Fe3O4-NPs mass 0.05g, pH 5 and initial concentration of Ce(III) ions 40mgL(-1). The SBA-15-BSEA-Fe3O4-NPs exhibited high adsorption efficiency and very good selectivity through cerium removal even in the presence of other ions (La(3+), Nb(3+), Er(3+), Cu(2+), Cd(2+), Cr(3+), and Fe(2+) ions). The SBA-15-BSEA-Fe3O4-NPs was successfully regenerated and the response was reversible. The R.S.D. of the adsorption process was less than 1.02%.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
569615
(3-Chloropropyl)triethoxysilane, 95%
C9H21ClO3Si