EMAIL THIS PAGE TO A FRIEND
Analytica chimica acta
An enhanced chemiluminescence resonance energy transfer aptasensor based on rolling circle amplification and WS2 nanosheet for Staphylococcus aureus detection.
PMID 28159108
Abstract
A chemiluminescence resonance energy transfer aptasensor was fabricated for the detection of Staphylococcus aureus (S. aureus) with Co(2+) enhanced N-(aminobutyl)-N-(ethylisoluminol) (ABEI) functional flowerlike gold nanoparticles (Co(2+)/ABEI-AuNFs) as donor and WS2 nanosheet as acceptor. In the presence of S. aureus, rolling circle amplification (RCA) can be started. Partially complementary sequence of RCA product functional ABEI-AuNFs (cDNA-ABEI-AuNFs) were then annealed to multiple sites of the RCA product to form duplex complex. This complex is less adsorbed onto the WS2 nanosheet, thus attenuating the quenching of ABEI-AuNFs chemiluminescence by WS2 nanosheet. In the absence of target S. aureus (and hence the absence of RCA and duplex formation), the free cDNA-ABEI-AuNFs is completely adsorbed onto the WS2 nanosheet and chemiluminescence quenching ensues. Under optimal conditions, the logarithmic correlation between the concentration of S. aureus and the CL signal was found to be linear within the range of 50xa0cfu/mL to 1.5xa0×xa010(5)xa0cfu/mL (R(2)xa0=xa00.9913). The limits of detection of the developed method were found to be 15xa0cfu/mL for S. aureus. The selectivity and the capability of the biosensor in meat samples were also studied. Therefore, this simple and easy operation method can be used to detect S. aureus with high sensitivity and specificity.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
A0156
N-(4-Aminobutyl)-N-ethylisoluminol, ≥90%
C14H20N4O2