EMAIL THIS PAGE TO A FRIEND
Journal of ethnopharmacology
Vasorelaxation effect of Glycyrrhizae uralensis through the endothelium-dependent Pathway.
PMID 28161542
Abstract
Glycyrrhiza uralensis (G. uralensis) is one of the herbs used in traditional Chinese medicine (TCM) and serves as an envoy medicinal. Since G. uralensis plays a major role in the anti-hypertensive TCM formulae, we believe that G. uralensis might possess vasorelaxation activity. This study is designed to investigate the vasorelaxation effect of G. uralensis from various extracts and to study its pharmacology effect. The vasorelaxation effect of G. uralensis extracts were evaluated on thoracic aortic rings isolated from Sprague Dawley rats. Among these three extracts of G. uralensis, 50% ethanolic extract (EFG) showed the strongest vasorelaxation activity. EFG caused the relaxation of the aortic rings pre-contracted with phenylephrine either in the presence or absence of endothelium and pre-contracted with potassium chloride in endothelium-intact aortic ring. Nω-nitro-L-arginine methyl ester, methylene blue, or 1H-[1,2,4]Oxadiazolo[4,3-a]quinoxalin-1-one inhibit the vasorelaxation effect of EFG in the presence of endothelium. On the other hand, in the presence of the potassium channel blockers (tetraethylammonium and barium chloride), the vasorelaxation effect of EFG was not affected, but glibenclamide and 4-aminopyridine did inhibit the vasorelaxation effect of EFG. With indomethacin, atropine and propranolol, the vasorelaxation effect by EFG was significantly reduced. EFG was also found to be effective in reducing Ca(2+) release from sarcoplasmic reticulum and the blocking of calcium channels. The results obtained suggest that EFG is involved in the NO/sGC/cGMP pathway.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
O3636
1H-[1,2,4]Oxadiazolo[4,3-a]quinoxalin-1-one, powder
C9H5N3O2