EMAIL THIS PAGE TO A FRIEND
Current protocols in microbiology
Efficient Genome Editing in the Oomycete Phytophthora sojae Using CRISPR/Cas9.
PMID 28166383
Abstract
Phytophthora is a filamentous fungus-like microorganism, but belongs to the oomycetes, in the kingdom Stramenopila. Phytophthora species are notorious as plant destroyers, causing multibillion-dollar damage to agriculture and natural ecosystems worldwide annually. For a long time, genome editing has been unattainable in oomycetes, because of their extremely low rate of homologous recombination. The recent implementation of the CRISPR/Cas (clustered regularly interspaced short palindromic repeats/CRISPR-associated) system in the soybean pathogen Phytophthora sojae, an experimental model for oomycetes, has opened up a powerful new research capability for the oomycete community. Here, we describe a detailed protocol for CRISPR/Cas9-mediated genome editing in P. sojae, including single guide RNA (sgRNA) design and construction, efficient gene replacement, and mutant-screening strategies. This protocol should be generally applicable for most culturable oomycetes. We also describe an optimized transformation method that is useful for other Phytophthora spp. including P. capsici and P. parasitica. © 2017 by John Wiley & Sons, Inc.