EMAIL THIS PAGE TO A FRIEND
Romanian journal of morphology and embryology = Revue roumaine de morphologie et embryologie
Evaluation of cardiac microvasculature in patients with diffuse myocardial fibrosis.
PMID 28174803
Abstract
Myocardial fibrosis is one of the most common histopathological lesions found in chronic heart diseases. Progressive development of myocardial fibrosis will cause heart failure, an extremely debilitating and life threatening condition. The correlation between the severity of fibrosis and myocardial microcirculation is an important prognostic factor in this disease entity. In our study, myocardial microvascular density evaluation of the patients with high blood pressure (hypertension), atrial fibrillation (AF), coronary heart disease, and heart failure showed a significant decrease of the values of this parameter, which means that myocardial fibrosis is the direct result of stimulation of myocardial fibroblasts induced by local hypoxia.