EMAIL THIS PAGE TO A FRIEND
Biomedicine & pharmacotherapy = Biomedecine & pharmacotherapie
25-Hydroxyvitamin D-1-α-hydroxylase in apoliporotein E knockout mice: The role of protecting vascular smooth muscle cell from calcification.
PMID 28178628
Abstract
Previous publications widely reported that 25-hydroxyvitamin D-1-α-hydroxylase (CYP27B1) regulated the metabolism of 25-hydroxyvitamin D3, which has a close association between altered activity of vitamin D and vascular calcification has been reported in various human diseases, including chronic kidney disease, osteoporosis and atherosclerosis. Vascular calcification is a clinically significant component of atherosclerosis and may be promoted by ROS associated inflammatory. In this study, we evaluated the effect of 25-hydroxyvitamin D-1-α-hydroxylase on the atherosclerosis disease both in apolipoprotein (apo) E-/- mice and wild-type mice. We also isolated endothelial cell (ECs) and vascular smooth muscle cells (VSMCs) in aortic from the wild type mice and apoE-/- mice respectively, then investigated that after parathyroid hormone (PTH) both of the CYP27B1 and vitamin D receptor (VDR) expressions in apoE-/-EC and apoE-/-VSMC were higher than the wide-type EC and VSMCs. However, the increased proliferation and decreased apoptosis have showed in EC and VSMC compared with the cells from apo E-/- mice. Moreover, the index associated with vascular calcification such as intracellular Ca(2+) concentration and alkaline phosphatase (ALP) activity have been tested and the result suggested that the levels of the former index have improved in the apoE-/-EC and apoE-/-VSMC. We got similar conclusions under the pre-treatment with 1, 25(OH) 2D3.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
46393
Fluo-3, suitable for fluorescence, ~70%
C36H30Cl2N2O13
G5500
L-(−)-Glucose, ≥99%
C6H12O6