EMAIL THIS PAGE TO A FRIEND
Apoptosis : an international journal on programmed cell death
Activation of Na(+)-K(+)-ATPase with DRm217 attenuates oxidative stress-induced myocardial cell injury via closing Na(+)-K(+)-ATPase/Src/Ros amplifier.
PMID 28181111
Abstract
Reduced Na(+)-K(+)-ATPase activity has close relationship with cardiomyocyte death. Reactive oxygen species (ROS) also plays an important role in cardiac cell damage. It has been proved that Na(+)-K(+)-ATPase and ROS form a feed-forward amplifier. The aim of this study was to explore whether DRm217, a proved Na(+)/K(+)-ATPase's DR-region specific monoclonal antibody and direct activator, could disrupt Na(+)-K(+)-ATPase/ROS amplifier and protect cardiac cells from ROS-induced injury. We found that DRm217 protected myocardial cells against hydrogen peroxide (H2O2)-induced cardiac cell injury and mitochondrial dysfunction. DRm217 also alleviated the effect of H2O2 on inhibition of Na(+)-K(+)-ATPase activity, Na(+)-K(+)-ATPase cell surface expression, and Src phosphorylation. H2O2-treatment increased intracellular ROS, mitochondrial ROS and induced intracellular Ca(2+), mitochondrial Ca(2+) overload. DRm217 closed Na(+)-K(+)-ATPase/ROS amplifier, alleviated Ca(2+) accumulation and finally inhibited ROS and mitochondrial ROS generation. These novel results may help us to understand the important role of the Na(+)-K(+)-ATPase in oxidative stress and oxidative stress-related disease.
Related Materials