EMAIL THIS PAGE TO A FRIEND
Chemosphere
Effect of various selenium doses on chromium(IV)-induced nephrotoxicity in a male chicken model.
PMID 28183056
Abstract
Our study aimed to explore whether Na2SeO3 (Se) can alleviate the nephrotoxicity induced by K2Cr2O7 [Cr(VI)]. One hundred and five male chickens were randomly divided into seven groups with 15 chickens each group: The 6 experimental groups received K2Cr2O7 alone or in combination with 0.31, 0.63, 1.25, 2.50, and 5.00xa0mg/kg for 42 days, respectively, while control group was treated with equivalent water. Exposure to Cr(VI) significantly increased MDA contents and organ coefficient, whereas decreased T-SOD activities, Ca(2+)-ATPase activities, mitochondrial membrane potential and GSH contents, and histological studies demonstrated renal damage. Above indicators were restored by Se supplement (0.31, 0.63, and 1.25xa0mg/kg), in which supplement with 0.63xa0mg/kg Se developed more effectively than the other two groups; on the contrary, in the groups of Se supplement with 2.50 and 5.00xa0mg/kg, the above indicators were not ameliorated and even exacerbated. This study demonstrated that Cr(VI) can result in kidney oxidative damage in male chickens, and Se of certain dose has the protective effects against Cr(VI)-induced nephrptoxicity.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
T4069
JC-1, solid
C25H27Cl4IN4