EMAIL THIS PAGE TO A FRIEND
The Journal of biological chemistry
Characterization of genomic deletion efficiency mediated by clustered regularly interspaced short palindromic repeats (CRISPR)/Cas9 nuclease system in mammalian cells.
PMID 28188222