EMAIL THIS PAGE TO A FRIEND
European journal of pharmaceutical sciences : official journal of the European Federation for Pharmaceutical Sciences
Extensive preclinical evaluation of an infliximab biosimilar candidate.
PMID 28188909
Abstract
Infliximab is therapeutic monoclonal antibody (mAb) against TNF-α employed in the treatment of immunoinflammatory diseases. The development of biosimilar mAbs is a global strategy to increase drug accessibility and reduce therapy-associated costs. Herein we compared key physicochemical characteristics and biological activities produced by infliximab and infliximab-Probiomed in order to identify functionally relevant differences between the mAbs. Binding of infliximab-Probiomed to TNF-α was specific and had kinetics comparable to that of the reference product. Both mAbs had highly similar neutralizing efficacy in HUVEC cell cultures stimulated with TNF-α. In vitro induction of CDC and ADCC were also similar between the evaluated products. In vivo comparability was assessed using a transgenic mouse model of arthritis that expresses human TNF-α in a 13-week multiple-administration study. Infliximab and infliximab-Probiomed showed comparable efficacy, safety, and pharmacokinetic profiles. Our results indicate that infliximab-Probiomed has highly similar activities to infliximab in preclinical models, warranting a clinical evaluation of its biosimilarity.