EMAIL THIS PAGE TO A FRIEND
ChemMedChem
Trachycladines and Analogues: Synthesis and Evaluation of Anticancer Activity.
PMID 28195671
Abstract
The synthesis of four new analogues of marine nucleoside trachycladine A was accomplished by direct regio- and stereoselective Vorbrüggen glycosylations of 2,6-dichloropurine and 2-chloropurine with a d-ribose-derived chiron. Naturally occurring trachycladines A and B and a series of analogues were examined for their cytotoxic activity against a number of cancer cell lines (glioblastoma, lung, and cervical cancer). Parent trachycladine A and two analogues (the diacetate of the 2,6-dichloropurine derivative and N-cyclopropyl trachycladine A) resulted in a significant decrease in cell viability, with the latter exhibiting a stronger effect. The same compounds enhanced the cytotoxic effect of docetaxel in lung cancer cell lines, whereas additional experiments revealed that their mode of action relies on mitotic catastrophe rather than DNA damage. Moreover, their activity as autophagic flux blockers was postulated.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
861677
DL-Cysteine, technical grade
C3H7NO2S