EMAIL THIS PAGE TO A FRIEND
Journal of pharmaceutical and biomedical analysis
Comparison of bioactive components and pharmacological activities of ophiopogon japonicas extracts from different geographical origins.
PMID 28196345
Abstract
Ophiopogon japonicus (Linn. f.) Ker-Gawl (O. japonicas), mainly cultivated in Sichuan and Zhejiang province in China, has different bioactive components and therefore their pharmacological activities. To explain the different clinical efficacy of O. japonicas derived preparations, herein we report differences of pharmacological activities between Sichuan and Zhejiang O. japonicas and behind them the exact differences of bioactive components. Based on a LC/MS-IT-TOF method, the differences of bioactive components between Sichuan and Zhejiang O. japonicas extracts were analyzed and respective characteristic components were picked out. We determined 39 ophiopogonones and 71 ophiopogonins compounds in Sichuan and Zhejiang O. japonicas extracts and found the contents of these compositions have several times difference. Evidenced by experimental data of pharmacological activities in inhibiting cardiomyocyte damage induced by H2O2, mouse macrophage cell inflammation induced by lipopolysaccharide and cytotoxicity in vitro, Zhejiang O. japonicas extract had a stronger antioxidant and anti-inflammatory capacity than Sichuan O. japonicas extract, and the two O. japonicas extracts exhibited selective cytotoxicity on different cancer cell lines in vitro. These data shed light on the links between bioactive components and pharmacological activities of O. japonicas derived preparations. Thus, geographical origin of O. japonicas should be considered to be a key factor in efficacy studies and further clinical application.