EMAIL THIS PAGE TO A FRIEND
International journal of oncology
Repositioning of amprenavir as a novel extracellular signal-regulated kinase-2 inhibitor and apoptosis inducer in MCF-7 human breast cancer.
PMID 28197631
Abstract
Computational drug repositioning by virtually screening existing drugs for additional therapeutic usage could efficiently accelerate anticancer drug discovery. Herein, a library of 1447 Food and Drug Administration (FDA)-approved small molecule drugs was screened in silico for inhibitors of extracellular signal-regulated kinase 2 (ERK2). Then, in vitro kinase assay demonstrated amprenavir, a HIV-1 protease inhibitor, as a potential kinase inhibitor of ERK2. The in vivo kinase assay indicated that amprenavir could inhibit ERK2-mediated phosphorylation of BimEL at Ser69. Amprenavir could suppress this phosphorylation in MCF-7 cells, which may further facilitate the association of BimEL with several pro-survival molecules. Additionally, inhibition of ERK2-BimEL signaling pathway by amprenavir could contribute to its anti-proliferative and apoptosis-inducing activity in MCF-7 cells. Finally, in vivo tumor growth and immunohistochemical studies confirmed that amprenavir remarkably suppressed tumor proliferation and induce apoptosis in MCF-7 xenografts. Taken together, amprenavir can effectively inhibit the kinase activity of ERK2, and thus induces apoptosis and inhibits tumor growth in human MCF-7 cancer cells both in vitro and in vivo, making amprenavir a promising candidate for future anticancer therapeutics.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
SML0685 Amprenavir, ≥98% (HPLC)
C25H35N3O6S