EMAIL THIS PAGE TO A FRIEND
Journal of experimental botany
Expression of cyanobacterial FBP/SBPase in soybean prevents yield depression under future climate conditions.
PMID 28204603
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
B6916 Bradford Reagent, for 0.1-1.4 mg/ml protein