EMAIL THIS PAGE TO A FRIEND
Physical therapy
Defining Patterns of Sagittal Standing Posture in Girls and Boys of School Age.
PMID 28204737
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
SML0463
BPD, ≥98% (HPLC)
C17H10O5